VWR® Measuring Cylinder, Hexagonal Base, with Stopper, Class A, Unserialized
Supplier: VWR, part of Avantor
VWR®
142.432.01A 142.432.06A 142.402.07A 142.402.05A 142.432.04A 142.402.03A 142.432.03A 142.402.02A 142.402.06A 142.432.05A 142.432.07A 142.432.02A 142.402.01A 142.402.08A 142.432.08A 142.402.04A
76004-056CS 391.9 USD
76004-056 76004-058 76003-716 76003-748 76003-726 76003-714 76003-724 76003-746 76004-060 76004-052 76003-718 76004-054 76003-728 76003-750 76003-722 76003-720
VWR® Measuring Cylinder, Hexagonal Base, with Stopper, Class A, Unserialized
Cylinders Measuring Cylinders
VWR® Meausuring Cylinders are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Hexagonal base
  • Calibrated 'to Deliver'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring Cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certificaciones: Cylinders are supplied with Batch certificates.