VWR® Measuring Cylinder, Hexagonal Base, TD, with Spout, Class A, Unserialized
Supplier: VWR, part of Avantor
VWR®
139.464.02A 139.464.05A 139.464.07A 139.464.06A 139.464.08A 139.464.04A 139.464.03A 139.464.01A
76003-696CS 260.78 USD
76003-696 76003-686 76003-694 76003-684 76003-688 76003-692 76003-682 76003-690
VWR® Measuring Cylinder, Hexagonal Base, TD, with Spout, Class A, Unserialized
Cylinders Measuring Cylinders
VWR® Measuring Cylinders are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Hexagonal base
  • Calibrated 'to Deliver'
  • Cylinders are made to meet ASTM E 1272 norms
  • Printed double scale
VWR® Measuring Cylinders are used for the precise measurement liquid volumes. Cylinders have a large hexagonal base to increase stability. Calibration and tolerances are in compliance to standards: ASTM E1272, ASTM E542, and ASTM E694.
Certificaciones: Cylinders are supplied with Batch certificates.