VWR® Graduated Pipettes (Mohr), Class A, Serialized
Supplier: VWR, part of Avantor
VWR®
125.452.03 125.452.01A 125.452.05 125.452.02 125.452.04 125.452.01
76003-564CS 220.94 USD
76003-564 76003-562 76003-568 76003-566 76003-560 76003-570
VWR® Graduated Pipettes (Mohr), Class A, Serialized
Pipets Measuring (Mohr) Pipets
VWR® pipettes are manufactured from ASTM E 438, Type 1, Class A Borosilicate 3.3 glass.
  • Calibrated to deliver at 20 °C per ASTM E 1293 according to Class A norms
VWR® Graduated Pipettes (Mohr) are calibrated to deliver a precise volume at a particular temperature. They are used for accurate measurements of liquid volumes. Calibration and toleration standards are defined as per standard specifications and practices: ASTM E1044, ASTM E1293, ASTM E542, and ASTM E694.
Certificaciones: Pipettes are supplied with individual work certificates.