VWR® Cylinders with Single Metric Scale and Glass Penny Head Stopper, Class B
Supplier: VWR, part of Avantor
VWR®
142.434.06A 142.434.05A 142.434.03A 142.434.02A 142.434.04A 142.434.08A 142.434.07A 142.434.01A
75997-932CS 359.59 USD
75997-932 75997-924 75997-934 75997-930 75997-926 75997-936 75997-928 75997-938
VWR® Cylinders with Single Metric Scale and Glass Penny Head Stopper, Class B
Cylinders Measuring Cylinders
VWR® Measuring Cylinders are made from Borosilicate 3.3 glass, ASTM 438, Type 1, Class A standard
Cylinders have a large hexagonal base to increase stability. The cylinder’s mouth is equipped with a joint to accommodate a glass stopper. Calibration and toleration standards for measuring cylinders are defined in the standard specifications and practices: ASTM E1272, ASTM E542, ASTM E694, and ISO 4788.
These cylinders are calibrated 'To Contain' and 'To Deliver' precise volumes at a particular temperature, and may be used to measure liquid volume.