VWR® Storage Carts
Supplier: VWR, part of Avantor
90818 90791 90802 90808 90804 90801 90642 90810 90821 90819 90799 90847 90828 90809 90788 90824 90786 90855 90846 90811 90822 90641 90795 90839 90817 90792 90644 90640 90837 90790 90832 90848 90850 90784 90845 90800 90816 90794 90643 90785 90823 90806 90797 90840 90833 90793 90807 90814 90805 90798 90825
75857-098EA 2707.15 USD
75857-098 75857-132 75857-092 75857-094 10809-056 75857-090 10809-054 10809-052 10809-050 75870-360 75870-364 75857-126 75870-400 75857-128 75870-366 75857-122 75857-124 75857-086 75857-088 75857-084 75857-080 75870-374 75870-378 75857-118 75857-078 75857-112 75857-074 75857-076 75857-110 75857-070 75857-072 75870-382 75870-380 75857-108 75870-342 75870-384 75857-104 75857-106 75870-388 75870-344 75857-100 75857-102 75857-066 75857-060 10809-048 75857-062 75870-352 75870-350 75870-394 75857-134 75857-058
VWR® Storage Carts
Carts
Storage carts made from food-grade, high-density polyethylene have four 3" industrial casters, two of which lock.
  • Locking hinged door
  • Bulk storage area with door
  • One key locks drawers and bulk storage
  • Full-extension drawers with handle
  • Polyethylene construction resists chemicals
The carts feature full-extension drawers for full viewing of drawer contents
The carts add extra storage and work space in labs, clinics, hospitals, and other locations. They move all supplies together in a single work area, so no one needs to make extra trips to set up an activity.
They create storage for glassware, chemicals, pens, pipets, tools, tweezers, bottles, and other supplies. The top-side shelves are a built-in dispenser for Kimwipes and gloves. Shelves snap in and out as needed. All drawers and bulk storage areas lock with provided keys.
Información para pedidos: Five flexible storage carts include 1, 2, 3, or 4 full-extension drawers, 1 large bulk storage area, 4 or 6 side shelves, and 2 built-in dispensers for gloves and wipes. The carts are available in five sizes and multiple colors. A total-replacement lifetime manufacturer's guarantee is provided. A rail top for security is optional.