VWR® Powerpette® Plus Pipet Controller
Supplier: VWR, part of Avantor
Powerpette®
612-3686 613-4448 613-4446 613-4445 613-4442 613-4447
75856-452EA 398.35 USD
75856-452 75856-450 75856-448 75856-446 75856-456 75856-454
VWR® Powerpette® Plus Pipet Controller
Pipets Pipet Controllers
The Powerpette plus pipet controller is a compact and portable device that features multiple user-selectable modes and speeds to perform a wide range of liquid handling applications.
  • Compact, self-contained design
  • Designed for left-hand or right-hand use
  • Suitable for a wide range of liquid handling applications
  • Accommodates pipets ranging from 1 to 100 ml
This lightweight, rechargeable pipet controller is ideal for a wide range of liquid handling applications. Controller features user-selectable aspirating and dispensing speeds and can be used in either 'to deliver' or 'blowout' mode. Compact, self-contained design with precision molding and concave finger triggers for comfortable handling. Pistol-style grip and an angled pipet holder nozzle allow use in small spaces such as laminar flow hoods or biological safety cabinets. Bench stand enables unit to be parked in upright position or with a pipet fitted. Suitable for large or small hands and for both left- and right-handed users. Works with pipets ranging from 1 to 100 ml.
The Powerpette® Plus Pipet Controller is supplied with a 3-year material and workmanship warranty, effective from the date of delivery.
Certificaciones: CE marked.
Información para pedidos: Unit is fitted with a 0.45 µm filter. Also supplied with manufacturer’s 3-year warranty, one 0.45 µm and one 0.2 µm hydrophobic filter, charger, and bench stand.