VWR® Safetypette™ Pipet Filler
Supplier: VWR, part of Avantor
Safetypette™
JENC266-003 612-4548 JENC266-004
14526-208EA 4.95 USD
14526-208 75856-442 14526-214
VWR® Safetypette™ Pipet Filler
Pipets Pipet Controllers
Designed to reduce the strain associated with serial pipetting while providing simple, precise operation for right- and left-handed users.
  • Reduces strain
  • Ambidextrous
  • Lightweight
Compatible with 1 to 100 ml blow-out and 'to deliver' pipets.
As much as 65 ml can be aspirated from a single squeeze of the bulb. Aspiration and dispensation speed is precisely controlled with the simple thumb lever. A mini squeeze bulb evacuates blow out pipets. Silicone pipet holder is washable and autoclavable.
Weighs only 102 g, sits comfortably in either hand.
Información para pedidos: Supplied with replaceable integral 0.45 µm filter and mint, white, pink, and mauve cones for color coding. Three-year manufacturer's warranty also included.