Lysing Matrix B, MP Biomedicals
Supplier: MP Biomedicals
116981010 116540425 116911050 116971050 116981001 116540429 116971025 116911100 116911500 116971100
75856-174EA 1368.69 USD
75856-174 75856-172 75856-150 75856-406 75856-404 75784-616 75784-614 75856-148 75856-146 75784-618
Lysing Matrix B, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix B tubes are used primarily for lysis of gram positive and gram negative bacteria, and will also disrupt fungal tissue and spores
  • Impact resistant tube containing 0.1mm silica spheres
  • Can be used as an additive for disruption by sonication
  • Use for gram-positive and gram-negative bacteria
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.