Lysing Matrix D, MP Biomedicals
Supplier: MP Biomedicals
116983010 116913050 116913500 116540434 116933050 116973025 116983001 116973050 116913100
75856-176EA 204.28 USD
75856-176 75856-154 75856-152 IC116933050 IC116913100 IC116540434 75784-626 75856-178 75784-606
Lysing Matrix D, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing matrix D is for rapid and reliable biological sample lysis from a wide variety of starting materials.
  • Impact resistant tube containing 1.4mm ceramic spheres
  • Primarily used for the lysis of softer tissues
  • Will also lyse plant tissue, cultured cells and whole insects
  • Grinding matrix is chemically inert and will not bind nucleic acids
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.