VWR® Standard Dry Block Heaters, 230 V (Export Only)

Supplier: VWR, part of Avantor
VWR®
80863274 80863252 80863258 80863268
75838-304EA 1571.09 USD
75838-304 75838-268 75838-272 75838-292
VWR® Standard Dry Block Heaters, 230 V (Export Only)
Block Heaters Analog Block Heaters
Multi-purpose, high wattage, constant temperature VWR Standard Dry Block heaters are economical, versatile and compact.

  • New design with smaller footprint
  • Designed for Multi-Purpose Use
  • Exceptional Temperature Uniformity
  • Holds Interchangeable Modular Heating Blocks
  • Heats to 150°C (302°F)
  • 5-Year-Warranty on Parts & Labor
  • Manufactured in the USA

Multi-purpose units are ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points, and a variety of other laboratory procedures

Each model accepts separate interchangeable modular blocks, which accommodate various sample enclosures such as vials, microplates, PCR strips, and a wide variety of tubes. Each block contains a thermometer well, and features close tube-and-block contact for maximum heat retention.

All units offer two separate temperature adjustment knobs providing reproducible low temperature and high temperature control. Low range knob adjusts from ambient to 100°C (212°F) and high range knob adjusts from 75 to 150°C (167 to 302°F).

Operating Conditions: Units can be run in conditions from 18 to 33°C (64 to 91°F) 20% to 80% relative humidity, non-condensing.

Certificaciones: CE Marked, TUV listed, RoHS Compliant.

Información accesorios: Heaters require a VWR modular heating block for operation (sold separately). Over 40 modular heating block options are available.

Información para pedidos: Unit must be ordered with heating block(s).

All units include a 234cm (92") detachable, Euro style plug.

Precaución: To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR