VWR® Repeating Dispenser and Syringe Tips
Supplier: VWR, part of Avantor
VWR®
ES42020-0000V ES43001-0147V ES40007-0009V ES43001-0148V ES40007-0020V ES40007-0010V ES40007-0019V
75836-686EA 861.34 USD
75836-686 75836-688 75838-210 75838-226 75836-690 75838-228 75838-208
VWR® Repeating Dispenser and Syringe Tips
Pipets Repetitive Pipettors
Designed for quick, accurate, and easy repeat pipetting. The smooth-moving dosing button and lightweight design facilitates long dosing series without causing strain or discomfort to the user.
  • Lightweight, ergonomic design
  • Allows for 9 tip sizes, 10 volume adjustments, and 90 program steps
Nine different tip sizes are available, for dispensing volumes ranging from 1 μl to 50 ml. The device provides reliable and repeatable results for a wide range of liquids, including viscous, infectious, and volatile samples. Available in sterile and nonsterile options. All sterile tips are individually wrapped.
Volume selection, loading, and dispensing of liquids can be performed with ease using only one hand.
Certificaciones: All sterile tips are certified to be pyrogen-, ATP-, RNase-, and DNase-free. The VWR® Repeating Dispenser is manufactured under DIN EN ISO 8655 requirements using high-quality materials, ensuring a device that is built to last.