VWR® Drosophila Vials, Narrow and Wide
Supplier: VWR, part of Avantor
3810 3813 3812 3811 3816 3815 3807 3814 3820 3819 3808 3809
75813-144CS 51.32 USD
75813-144 75813-166 75813-156 75813-146 75813-158 75813-148 75813-160 75813-150 75813-162 75813-152 75813-164 75813-154
VWR® Drosophila Vials, Narrow and Wide
Vials
Variety of sizes and materials for all applications.
  • High quality, virgin grade plastic
  • Wide and narrow opening diameters
  • Tray packs enable easier food loading
VWR® Drosophila vials provide multiple options for storing fly cultures. Manufactured from pure virgin medical grade plastics, the vials are available in crystal clear polystyrene, translucent flexible and autoclavable polypropylene, or virtually unbreakable K-Resin. The flat bottom, straight wall design allows for convenient racking, stacking, and filling. The vials are available in bulk quantities, or in a 10 by 10 tray pack for quick and effortless filling. The bulk quantities are layered with open-end orientation for convenient handling and small case size.