EZwaste™ Safety Tray Secondary Containers, Foxx Life Sciences
Supplier: Foxx Life Sciences
252-1903-OEM 252-1901-OEM 252-1907-OEM 252-1902-OEM 252-1905-OEM 252-1906-OEM 252-1908-OEM 252-1904-OEM
75805-324EA 108.52 USD
75805-324 75805-476 75805-332 75805-478 75805-328 75805-330 75805-334 75805-326
EZwaste™ Safety Tray Secondary Containers, Foxx Life Sciences
Trays/Dishes/Pots Spillage Trays/Pans
An ideal solution for preventing leakage and spillage during carboy storage or transport.
  • Tough and durable
  • Designed with the capacity to hold 115% of carboy content
EZwaste™ safety tray secondary containers are a perfect solution for preventing leakage and spillage from EZwaste™ solvent waste system containers.