Lysing Matrix E, MP Biomedicals
Supplier: MP Biomedicals
116914050 116984001 116914100 116914500 116984010
75784-630PK 348.86 USD
75784-630 75856-384 IC116914500 75856-180 75784-628
Lysing Matrix E, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix E is effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumors and other difficult tissues.
  • Impact resistant tube
  • Contains 1.4mm ceramic spheres, 0.1mm silica spheres and 4mm glass beads
  • DNase and RNase Free
Lysing Matrices are critical components of the FastPrep™ Sample Preparation system. MP Biomedicals recieves the matrix particles from proven vendors and ensures the unprocessed material meets high qualitative standards. Matrix particles are then prepared and dispensed under a rigorous set of proprietary conditions that offer customers a premium product that can be immediately used with confidence.Matrices are available separately for use with your own unique buffers, and are also available as components of the complete purification kits.