Lysing Matrix C, MP Biomedicals
Supplier: MP Biomedicals
116912500 116912100 116912050
75784-622PK 277.44 USD
75784-622 75784-620 75784-624
Lysing Matrix C, MP Biomedicals
Homogenizers Benchtop Homogenizers
Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
  • Lysis of yeast and fungi
  • Used for molds, spores and filamentous algae
Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae
Each impact-resistant 2 mL tube contains 1 mm silica spheres. Lysing Matrix C tubes have red caps and are found in the FastRNA Pro Red Kit and FastProtein Red Matrix. Lysing Matrix C is used primarily for lysis of yeast and fungi and can also be used for molds, spores and filamentous algae.
This grinding matrix is chemically inert and will not bind nucleic acids.