SRK® Short Swabs, COPAN Diagnostics
Supplier: COPAN Diagnostics
SRK™
922C
75784-158CS 542.1 USD
75784-158
SRK® Short Swabs, COPAN Diagnostics
Swabs Swabs with Rinse Solutions
Swab rinse kits (SRK®) for microbiological examination of surfaces.
  • Isotonic salt solution to neutralize and deactivate disinfectants and sanitising agents
  • Rinse solution to moisten the sterile swab before sampling and transport it after collection
  • Swab shafts (rayon tip) are attached to the tube caps
Suitable for food and industrial applications.
Información de suministro: 50 swabs per box except the « bulk » item.