SP Bel-Art Buchner Table-Top Funnels, 260 mm (10.25"), Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a part of SP
H14620-0000 H14625-3510 H14620-1260 H14626-3510 H91463-1260 H91462-4026 H91462-4326 H14627-0000 H14627-1260
74950-378EA 361.61 USD
74950-378 13197-326 30308-090 74950-366 47750-812 47750-804 74950-362 47750-796 47750-786
SP Bel-Art Buchner Table-Top Funnels, 260 mm (10.25"), Bel-Art Products, a part of SP
Funnels Filtration Funnels Büchner Funnels
Filter large volumes quickly with a polyethylene plastic Table-Top Buchner Funnel using your choice of Fritware® or Perforated filter plate.
  • Removable hose barbed connector accepts 12.7 mm (½") I.D. tube and threads into ¾" NPT bung on the funnel
  • Overall height: 220 mm (8.75")
  • Rim to plate: 152 mm (6")
Fritware® Porous Filter Plates supported by a multiple ring grid, are made of 6.4 mm (¼") thick HDPE, with a non-porous ring at the periphery of the plate which seals filter paper when used. Fritware® Porous Filter Plates (fixed or removable) are available in two porosities (medium, 45 - 90 µm and coarse, 90 - 130 µm). Temperature range: –20...+52 °C (–4...125 °F).
Perforated Filter Plates are made of 4.8 mm (³/₁₆") thick HDPE with 4.8 mm (³/₁₆") perforations on 11 mm (⁷/₁₆") centers. Perforated Filter Plates (fixed or removable): Fixed for coarse filtration or use with a cloth or paper filter; Removable for use in precious metals recovery. Temperature range: –20...+52 °C (–4...125 °F).