SP Bel-Art Spinbar® Polygon, Magnetic Stirring Bars without Pivot Ring, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
F37120-0070 F37120-0025 F37120-0030 F37120-0012 F37120-0080 F37120-0040 F37120-0020 F37120-0002 F37120-0010 F37120-0038 F37120-0078 F37120-0112 F37120-0015 F37120-0003 F37120-7836 F37120-0060 F37120-0125 F37120-0050 F37120-0058 F37120-0212
74950-268EA 8.41 USD
74950-268 74950-266 74950-278 74950-264 74950-276 74950-262 47750-150 74950-274 74950-260 74950-272 74950-270 58947-185 58947-179 47750-152 47750-142 47750-154 47750-144 47750-156 47750-146 47750-148
SP Bel-Art Spinbar® Polygon, Magnetic Stirring Bars without Pivot Ring, Bel-Art Products, a part of SP
Stirrers Magnetic Stirring Bars
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
  • PTFE low friction coating
  • Durable and inert
  • Autoclave safe
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.