Micro Capillary Tubes, Disposable, Kimble Chase
Supplier: DWK Life Sciences
71900-25 71900-100 71900-20 71900-10
72690-008CS 367.69 USD
72690-008 72690-004 72690-006 72690-002
Micro Capillary Tubes, Disposable, Kimble Chase
Pipets Microcapillaries
Capillary action draws the fluid into the tube. To dispense, simply cover the hole on the rubber bulb and squeeze. Pipets are 5" long with a single black capacity ring. They are color-coded for selection of the correct size.
  • Packaged in easy-open cylinders for convenient dispensing
  • Clean and ready to use
  • Filling is easy
Certificaciones: Micro Capillary pipets made of soda lime glass in compliance with ASTM Specification E438, Type II glass requirements.
Envase: Packaging is designed from ASTM Specification E672 requirements. Each cylinder contains 250 pipets and one pipet device.