VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Supplier: VWR, part of Avantor
VWR®
66009-852A 89524-190A 89239-026A 66009-860A 66009-862A 82030-972A 10803-886A 10803-884A 10804-054A 10781-890A 82030-976A 66009-856A 89239-028A 46610-722A 89239-024A 46610-724A 10803-888A 46610-726A 82030-974A 89523-478A 89523-480A 89524-188A 82030-970A 76326-140A 10804-056A 66009-858A 66009-854A 89239-030A
46610-722ACS 340.58 USD
46610-722A 46610-724A 46610-726A 66009-862A 66009-852A 66009-860A 89239-030A 10803-888A 10803-884A 10803-886A 89239-026A 89239-028A 89239-024A 89524-190A 82030-976A 82030-972A 82030-974A 76326-140A 82030-970A 89523-480A 66009-858A 66009-856A 89524-188A 66009-854A 89523-478A 10781-890A 10804-056A 10804-054A
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
Vials
9 mm autosampler screw thread vials feature a wide opening design for compatibility with all major autosampler brands and robotics.
  • Vials are precision manufactured from polypropylene or high-purity borosilicate glass
  • Caps and vials matched for best closure integrity
  • Available with or without writing patch
  • Spring promotes alignment and ensures insert seals against septa to help guard against evaporation loss
  • Available pre-assembled, certified, clear or amber glass
MS certified vial kits are tested and certified to meet or exceed up to 14 critical performance parameters. Glass vials are manufactured from borosilicate glass and have a maximum fill volume of 1.8 ml.
Kits include preassembled polypropylene screw closures with your choice of PTFE/red rubber, PTFE/silicone/PTFE, or PTFE/silicone septa. Preassembled kits are supplied in one-compartment trays. Unassembled kits are supplied in two-compartment benchtop storage trays. Loose caps and inserts come packaged in poly bags.
Información para pedidos: Purchase vial and caps in convenience kit or separately.