VWR® Single Scale Graduated Cylinders, Polypropylene
Supplier: VWR, part of Avantor
VWR®
28457-6000 28455-6000 28452-6000 28456-6000 28454-6000 28453-6000 28458-6000
65000-006CS 203.91 USD
65000-006 65000-004 65000-008 65000-010 65000-002 65000-012 65000-000
VWR® Single Scale Graduated Cylinders, Polypropylene
Cylinders Measuring Cylinders
The “nonwetting” polypropylene (PP) plastic surface of these single-scale graduated cylinders eliminates concave meniscus and makes reading easier and more accurate.
  • Polypropylene construction
  • Chemically resistant
  • Autoclavable
  • Single metric scale has raised numerals for easy reading
  • Flared top rims and large spouts facilitate pouring
  • Octagonal-shaped bases prevent tipping and rolling
Chemically resistant, cylinders are autoclavable at 121°C (250°F).