SP Bel-Art Poxygrid® Half-Size Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
Poxygrid®
F18788-2000 F18788-1600 F18788-2001 F18788-1301 F18788-2000 F18788-2003 F18788-1603 F18788-1601 F18788-1300
60941-583CS 1683.5 USD
60941-583 60941-572 60941-594 60941-561 60941-619 60941-608 60941-621 490012-480 60941-596
SP Bel-Art Poxygrid® Half-Size Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
Excellent space-savers for use in water baths, laminar flow hoods, fume hoods or any place where space is limited.
  • Compact racks
  • Epoxy-coated steel wire construction
  • Chemical and corrosion-resistant
  • Unaffected by organic solvents, salts, weak acids, and weak alkalis
  • Autoclavable
These compact racks have square holes, three decks, slotted mesh bottom deck, and reinforced edges.