SP Bel-Art Poxygrid® 'Rack And A Half' Test Tube Racks, Bel-Art Products, a part of SP
Supplier: Bel-Art
F18786-0770 F18786-0780 F18786-0750
60916-609CS 913.85 USD
60916-609 60916-606 60916-600
SP Bel-Art Poxygrid® 'Rack And A Half' Test Tube Racks, Bel-Art Products, a part of SP
Racks Tube Racks
High-quality epoxy-coated wire racks in convenient extra-long sizes for volume test tube work.
  • Resistant to chemicals and corrosion
  • Autoclavable
  • Epoxy-coated steel wire construction
  • Conveniently sized
Epoxy-coated steel wire is chemical and corrosion resistant and steam autoclavable at 121°C (250°F).