VWR® Safe-T-Flex Caps
Supplier: VWR, part of Avantor
VWR®
6575 6530 6565 6510 6710 6505 6555 6525 6730 6630 6520 6500 6725 6715 6615 6515 6620 6720 6560 6570 6580 6600 6550 6605 6625 6610 6700 6705
60828-770PK 224.06 USD
60828-770 60828-772 60828-750 60828-730 60828-710 60828-732 60828-712 60828-738 60828-760 60828-740 60828-762 60828-764 60828-720 60828-742 60828-722 60828-700 60828-766 60828-744 60828-702 60828-746 60828-768 60828-724 60828-726 60828-704 60828-748 60828-706 60828-728 60828-708
VWR® Safe-T-Flex Caps
Closures Test/Sample Tube Closures
Safe-T-Flex caps feature a unique, flexible, over-locking design that locks securely on the outside of the tube lip and can be removed with one hand.
  • Polyethylene construction
  • Liquid-tight seal
  • Color-coded for easy specimen identification
  • Cap top can be labeled
  • Self-locking caps hold firmly in any position
A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples
Caps fit all varieties of blood collecting and disposable culture tubes. They are color-coded for easy specimen identification and to help guard against cross-contamination. Top of cap can also be numbered or labelled with indelible ink. Self-locking caps hold firmly in any position and keep contents secure while travelling. Liquid-tight seal withstands centrifugation, agitation, refrigeration, and freezing. Manufactured from polyethylene.
This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus.