VWR® Plug-Type Caps, Polyethylene
Supplier: VWR, part of Avantor
VWR®
3302-807-000 3302-804-000 3302-870-000 3302-801-000 3302-808-000 3302-802-000 3302-800-000
60819-047CS 406.21 USD
60819-047 60819-036 60819-069 60819-014 60819-025 60819-003 60819-080
VWR® Plug-Type Caps, Polyethylene
Closures Test/Sample Tube Closures
Disposable caps are for 12 x 75 mm and 13 x 100 mm culture tubes.
  • Disposable
  • Polyethylene construction
  • Unique fluted design
  • Snugly grasps plastic or glass tubes
They feature a unique fluted design that grips plastic or glass tubes snugly.
Información para pedidos: The assorted pack contains 500 caps each in yellow, orange, red, blue, green and purple.