VWR® Rubber Stoppers
Supplier: VWR, part of Avantor
VWR®
TW-ASSORT TW-9 TW-7 TW-10 TW-3 TW-5 TW-8 TW-4 TW-6
59586-157CS 741.79 USD
59586-157 59586-167 59586-159 59586-158 59586-164 59586-163 59586-160 59586-162 59586-161
VWR® Rubber Stoppers
Stoppers Rubber Stoppers
Versatile rubber stoppers are four stoppers in one.
  • Easy conversion between stopper types
  • Versatile design
  • Soft and resilient
  • Natural rubber construction
  • Autoclavable
Three simple twist-off seals permit easy conversion from a solid to a one, two, or three-hole stopper for all your laboratory needs. Soft, resilient, natural, white rubber demonstrates exceptional aging characteristics and withstands repeated autoclaving.
Información para pedidos: Stoppers are available in sizes 3 to 10 or as an assortment of 33 stoppers. Sizes 3 and 4 only have two twist-off seals. Stopper assortment 59586-167 contains nine size 3 stoppers, eight size 4, six size 5, four size 6, two size 7, two size 8, one size 9, and one size 10 stopper. For black stoppers (shown above), contact your Avantor representative.