VWR® Black Rubber Stoppers, Solid, without Hole
Supplier: VWR, part of Avantor
VWR®
7.5M290 13-M290 135M290 B00-M290 15-M290 9.5M290 8.5M290 9--M290 4--M290 14-M290 115M290 3--M290 12-M290 6.5M290 6--M290 0-M290 K8-M290 000M290 105M290 5--M290 7-M290 1--M290 11-M290 10-M290 2--M290 5.5M290
59580-080CS 298.13 USD
59580-080 59580-160 59580-182 59580-284 59580-240 59580-262 59580-364 59580-342 59580-386 59580-069 59580-322 59580-047 59580-488 59580-444 59580-466 59580-400 59580-422 59580-502 59580-546 59580-524 59580-149 59580-105 59580-127 59580-229 59580-207 59580-309
VWR® Black Rubber Stoppers, Solid, without Hole
Stoppers Rubber Stoppers
These solid, black stoppers contain a high percentage of pure rubber, weigh less than neoprene, and are low in free sulfur (1/2%).
  • Resilient and pliable for a long time
  • Low in free sulfur
  • Can withstand adverse laboratory conditions without hardening
Composition ensures resiliency and pliability over a long period.