VWR® 8" Test Sieves, All Stainless Steel
Supplier: VWR, part of Avantor
VWR®
.265"SS8H 1/8"SS8F 7/8"SS8H 325SS8F 16SS8F 5/8"SS8H 140SS8H 80SS8F 40SS8F 70SS8F 45SS8F 5"SS8F 500SS8F 25SS8H 45SS8H 20SS8F 3/4"SS8H 80SS8H 270SS8F 7SS8F 30SS8F 3.5SS8F 6SS8F 7/8"SS8F 5/16"SS8H 14SS8H 30SS8H 2-1/2"SS8F 3-1/2"SS8F 1-3/4"SS8F 400SS8H 3/4"SS8F 1/2"SS8F 400SS8F 10SS8H 6SS8H 4"SS8F 5SS8H 60SS8H 2.12"SS8H 200SS8F 1-1/2"SS8F 3/8"SS8F 7/16"SS8F 1/4"SS8H 10SS8F 2"SS8F 100SS8H 20SS8H 3.5SS8H 70SS8H 4.24"SS8F 14SS8F 7/16"SS8H 7SS8H 1.00"SS8H 18SS8H 12SS8H 120SS8H 450SS8F 140SS8F 230SS8H 120SS8F 60SS8F 1/8"SS8H .530"SS8F 5SS8F 450SS8H 1/2"SS8H 35SS8F 25SS8F 50SS8H 500SS8H 5/8"SS8F 1.00"SS8F 100SS8F 8SS8H 170SS8H 635SS8H 40SS8H 2.12"SS8F 3/8"SS8H 16SS8H 230SS8F 1/4"SS8F 12SS8F 635SS8F 325SS8H 50SS8F 1.06"SS8F 35SS8H 4SS8F .265"SS8F 200SS8H 8SS8F 18SS8F 4SS8H 1-1/4"SS8F 170SS8F 5/16"SS8F 270SS8H
57334-570EA 203.88 USD
57334-570 57334-452 57334-574 57334-450 57334-572 57334-456 57334-578 57334-454 57334-576 57334-458 57334-562 57334-442 57334-560 57334-440 57334-566 57334-446 57334-600 57334-564 57334-444 57334-604 57334-568 57334-448 57334-602 57334-394 57334-392 57334-430 57334-398 57334-552 57334-396 57334-550 57334-434 57334-556 57334-432 57334-554 57334-438 57334-436 57334-558 57334-418 57334-386 57334-540 57334-420 57334-387 57334-385 57334-544 57334-424 57334-388 57334-542 57334-422 57334-548 57334-428 57334-546 57334-426 57334-528 57334-408 57334-492 57334-490 57334-530 57334-494 57334-412 57334-534 57334-410 57334-532 57334-416 57334-538 57334-414 57334-536 57334-518 57334-482 57334-480 57334-486 57334-484 57334-522 57334-402 57334-520 57334-488 57334-400 57334-526 57334-406 57334-404 57334-470 57334-592 57334-590 57334-474 57334-596 57334-472 57334-594 57334-478 57334-476 57334-598 57334-516 57334-580 57334-460 57334-584 57334-464 57334-582 57334-462 57334-588 57334-468 57334-586 57334-466 57334-504
VWR® 8" Test Sieves, All Stainless Steel
Sieves Test Sieves
VWR 8" diameter (203.2 mm) all stainless steel test sieves are durable and ideal for sieving applications where a medium sample size is required.
  • Highly durable construction
  • Stainless steel fabrication
Each sieve bears a traceable serial number and comes with a certificate of compliance verifying your sieve was made in compliance to the current ASTM E11 and ISO 3310 specifications. Sieve pans, sieve covers, sieve brushes and Centerline® sieve inspection or calibration certificates are all available accessories.
Información accesorios: Half height (1") or full height (2") pans are available in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available in a variety of options, with brush head coarseness and handle design and construction being major decision factors.
Información para pedidos: Sieves are available in half height (1"), or full height (2"). Sieve height is measured from the frame rim to the sieve cloth.