VWR® High-Purity Two-Stage Gas Regulators, Stainless Steel
Supplier: VWR, part of Avantor
VWR®
3300768 3300771 3300763 3300773 3300764 3300770 3300769 3300774 3300765 3300767 3300772 3300766
55850-684EA 2167.22 USD
55850-684 55850-690 55850-674 55850-694 55850-676 55850-688 55850-686 55850-696 55850-678 55850-682 55850-692 55850-680
VWR® High-Purity Two-Stage Gas Regulators, Stainless Steel
Gas Regulators
Regulators have a stainless steel barstock body and are suitable for corrosive gas applications, high-purity gas applications, research sample systems, process analyzers, gas chromatography, EPA protocol mixes, laser gas systems, and emission monitoring systems.
  • Suitable for high-purity applications
  • Two-stage design for constant outlet pressure
  • Stainless steel diaphragm prevents contaminations
  • One-piece PTFE encapsulated seat design with sintered filter
The two-stage design provides constant outlet pressure regardless of change in cylinder pressure. They may be used with corrosive gas applications. A stainless steel diaphragm eliminates contamination from diffusion or outgassing. A one-piece PTFE encapsulated seat design includes a sintered filter to protect the seat from particulate contamination. The 1x10-9cc/sec. inboard helium leak rate maintains gas purity levels. Front panel mountable.
Información para pedidos: Other delivery pressures and outlet connections are available; contact your VWR sales representative for information.