VWR® High-Purity Single-Stage Gas Regulators, Stainless Steel
Supplier: VWR, part of Avantor
VWR®
3001163 3001164 3001156 3001157 3001167 3001160 3001159 3001161 3001158 3001162 3001166
55850-670EA 1631.94 USD
55850-670 55850-660 55850-650 55850-662 55850-652 55850-664 55850-654 55850-666 55850-656 55850-668 55850-658
VWR® High-Purity Single-Stage Gas Regulators, Stainless Steel
Gas Regulators
Regulators have a stainless steel barstock body and are suitable for a variety of applications where a rise in delivery pressure from a full to an empty cylinder can be tolerated.
  • Stainless steel body
  • Stainless steel diaphragm
  • Sintered filter
  • PTFE encapsulated seat design
They may be used with corrosive gas applications
Other delivery pressures and outlet connections are available; contact your VWR sales representative for information.
They are also ideal for high-purity gas applications, research sample systems, process analyzers, gas chromatography, EPA protocol mixes, laser gas systems, and emission monitoring systems. A stainless steel diaphragm eliminates contamination from diffusion or outgassing. A one-piece PTFE encapsulated seat design includes a sintered filter to protect the seat from particulate contamination. The 1x10-9cc/sec. inboard helium leak rate maintains gas purity levels. Front or back panel mountable.