VWR® High-Purity Two-Stage Gas Regulators, Brass
Supplier: VWR, part of Avantor
VWR®
3300762 3300756 3300753 3300760 3300759 3300758 3300752 3300754 3300761 3300755 3300757
55850-642EA 1055.49 USD
55850-642 55850-630 55850-624 55850-640 55850-636 55850-634 55850-622 55850-626 55850-638 55850-628 55850-632
VWR® High-Purity Two-Stage Gas Regulators, Brass
Gas Regulators
VWR® High-Purity Two-Stage Gas Regulators have a brass barstock body and are suitable for high-purity applications, research sample systems, process analyzers, gas chromatography, EPA protocol mixes, laser gas systems, and emission monitoring systems.
  • Suitable for high-purity applications
  • Two-stage design for constant outlet pressure
  • Stainless steel diaphragm prevents contaminations
  • One-piece PTFE encapsulated seat design with sintered filter
The two-stage design provides constant outlet pressure regardless of change in cylinder pressure. A stainless steel diaphragm eliminates contamination from diffusion or outgassing. A one-piece PTFE encapsulated seat design includes a sintered filter to protect the seat from particulate contamination. The 1x10-9cc/sec. inboard helium leak rate maintains gas purity levels. Front panel mountable.
Información para pedidos: Other delivery pressures and outlet connections are available; contact your VWR® sales representative for information.