VWR® High-Purity Single-Stage Gas Regulators, Brass
Supplier: VWR, part of Avantor
VWR®
3001151 3001150 3001152 3001145 3001148 3001154 3001149 3001153 3001147 3001155 3001146
55850-604EA 844.42 USD
55850-604 55850-616 55850-606 55850-618 55850-608 55850-620 55850-610 55850-600 55850-612 55850-602 55850-614
VWR® High-Purity Single-Stage Gas Regulators, Brass
Gas Regulators
Regulators have a brass barstock body and are suitable for a variety of applications where pressure rise is not critical, such as high-purity gas applications, research sample systems, process analyzers, gas chromatography, EPA protocol mixes, laser gas systems, and emission monitoring systems.
  • Brass body
  • Stainless steel diaphragm
  • Sintered filter
  • PTFE seat design
A stainless steel diaphragm eliminates contamination from diffusion or outgassing
Note: For noncorrosive gases only.
A one-piece PTFE encapsulated seat design includes a sintered filter to protect the seat from particulate contamination. The 1x10-9cc/sec. inboard helium leak rate maintains gas purity levels. Front or back panel mountable.
Información para pedidos: Other delivery pressures and outlet connections are available; contact your VWR® sales representative for information.