VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Supplier: VWR, part of Avantor
VWR®
3302653 3302651 3302663 3302646 3302660 3302637 3302645 3302650 3302642 3302644 3302639 3302654 3302652 3302649 3302638 3302662 3302661 3302640 3302648 3302656 3302647 3302641 3302643 3302657 3302659 3302664
55850-460EA 669.1 USD
55850-460 55850-440 55850-462 55850-464 55850-442 55850-420 55850-444 55850-422 55850-446 55850-424 55850-468 55850-426 55850-418 55850-450 55850-430 55850-452 55850-432 55850-412 55850-434 55850-456 55850-458 55850-436 55850-414 55850-438 55850-416 55850-428
VWR® Multistage Gas Regulators with Stainless Steel Diaphragms
Gas Regulators
VWR® Multistage gas regulators with stainless steel diaphragms are designed for gas chromatography applications where a constant working pressure is important over a wide range of inlet pressures and flow rates.
  • Solderless construction for easy maintenance
  • Solid forged brass body contains two regulators
  • First stage reduces full cylinder pressure to 90%
  • Sintered inlet filter removes particulates, tank scale, and rust from gas stream
  • Stainless steel diaphragms
  • Supply pressure gauge: 4000 psig
VWR® Multistage Gas Regulators feature safe, accurate, and sensitive delivery of reduced pressure
Note: For noncorrosive gases only.
Solderless construction for easy maintenance. Solid forged brass body contains two regulators. First stage reduces full cylinder pressure to 90% for safety. Particulates, tank scale, and rust are eliminated from gas stream with sintered inlet filter. Assembly features include: high-pressure valve, low-pressure valve, intermediate pressure relief valve between the two stages, back cap vent holes, and bonnet vent holes. Stainless steel diaphragms. High and low pressure PTFE encapsulated seat. Regulator damage from contaminants is minimal.