VWR® Multistage Gas Regulators with Neoprene Diaphragms
Supplier: VWR, part of Avantor
VWR®
3302628 3302631 3302619 3302626 3302630 3302629 3302621 3302625 3302618 3302635 3302627 3302620 3302617 3302636 3302622 3302624 3302632 3302633 3302623
55850-390EA 600.52 USD
55850-390 55850-490 55850-480 55850-392 55850-492 55850-482 55850-494 55850-472 55850-396 55850-484 55850-496 55850-474 55850-398 55850-486 55850-388 55850-476 55850-498 55850-488 55850-478
VWR® Multistage Gas Regulators with Neoprene Diaphragms
Gas Regulators
VWR® Multistage Gas Regulators with Neoprene Diaphragms are designed for applications where a constant working pressure is important over a wide range of inlet pressures and flow rates.
  • Solderless construction for easy maintenance
  • Solid forged brass body contains two regulators
  • First stage reduces full cylinder pressure to 90%
  • Sintered inlet filter removes particulates, tank scale, and rust from gas stream
VWR® Multistage Gas Regulators feature safe, accurate, and sensitive delivery of reduced pressure
Note: For noncorrosive gases only.
Solderless construction for easy maintenance. Solid forged brass body contains two regulators. First stage reduces full cylinder pressure to 90% for safety. Particulates, tank scale, and rust are eliminated from gas stream with sintered inlet filter. Assembly features include: high-pressure valve, low-pressure valve, intermediate pressure relief valve between the two stages, back cap vent holes, bonnet vent holes, and diaphragm backup plate with burst hole. Neoprene diaphragms. External safety to 250psig. Regulator damage from contaminants is minimal.