VWR® Ultra-High Delivery Pressure Regulator
Supplier: VWR, part of Avantor
VWR®
3200300 3200302
55850-300EA 798.41 USD
55850-300 55850-305
VWR® Ultra-High Delivery Pressure Regulator
Gas Regulators
The Model GP 8700 is a single stage high pressure regulator that is designed to operate on high pressure cylinders.
  • Ergonomic knob for improved grip
  • Dual scale gauges (psi/bar)
  • 2½” brass gauges
  • High pressure/low volume capacity
One piece encapsulated valve design.
Certificaciones: UL; CGA E-4