Nalgene® Pipet Baskets, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
DS5241-0050 DS5241-0040 DS5241-0020
53596-065CS 366.52 USD
53596-065 53596-043 53596-021
Nalgene® Pipet Baskets, Thermo Scientific
Pipets Pipet Washers
High-density polyethylene basket used for soaking or washing pipets.
For use in 36322-042 series pipet jars. Resilient plastic basket with handle that cushions pipets. Perforated screen is recessed 19 mm (³/₄") into base for complete drainage. Basket Ext.Ø.: 13.3 cm (5 ¹/₄").