Nalgene® Pipet Washer-Rinsers, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
5245-0050 5245-0040
53595-020CS 3082.65 USD
53595-020 53595-041
Nalgene® Pipet Washer-Rinsers, Thermo Scientific
Pipets Pipet Washers
One-piece, high-density polyethylene washer-rinser is leakproof.
Water inlet at the top prevents back-siphoning of contaminated water. Operates at wide range of cycling speeds, with water input from 1.5 to 12 L per minute.
Información para pedidos: Includes inlet and outlet tubing ready for immediate installation. Compatible pipet basket is available separately (see Accessories tab below).