VWR® Microdispenser Replacement Kits
Supplier: VWR, part of Avantor
3-000-310-K 3-000-388-K 3-000-005-K 3-000-375-K 3-000-005-K 3-000-320-K 3-000-385-K 3-000-325-K 3-000-350-K
53506-910EA 35.39 USD
53506-910 53506-900 53506-922 53506-912 53506-924 53506-914 53506-926 490012-670 53506-928
VWR® Microdispenser Replacement Kits
Pipets Repetitive Pipettors
The 5, 10, and 20 µl VWR Microdispenser Replacement Kits each include 2 plungers, 2 O-rings, 1 collet, and 1 coil spring and barrel tube.
  • Available in a variety of sizes
Información de suministro: The 20 and 25, 50, 100, 100/200, and 250 µl VWR Microdispenser Replacement Kits each include 1 plunger, 2 O-rings, 1 collet, 3 fluoropolymer resin tips, and 1 coil spring and barrel tube.