VWR® Fast-Release Pipette Pump™ II Pipetting Devices
Supplier: VWR, part of Avantor
VWR® Pipette Pump™
F37911-1002 F37911-1025 F37911-1010
53502-255CS 403.46 USD
53502-255 53502-253 53502-251
VWR® Fast-Release Pipette Pump™ II Pipetting Devices
Pipets Pipet Controllers
Pipetting devices feature a rapid release valve for quick dispensing.
  • Color-coded for quick identification
  • Easily disassembled for cleaning
A knurled thumb wheel rotates to aspirate liquid to the desired volume, and may be rotated to allow gradual dispensing. Apply light pressure to the fast release lever to rapidly dispense samples. Devices feature a unique collar with stabilizing fingers to securely hold glass or plastic pipets. Sizes are color coded, and all devices may be easily disassembled for cleaning.