Pipet Aid® Portable Pipetting Device, Drummond Scientific
Supplier: Drummond
Pipet-Aid®
4-000-200 4-000-100
53498-104EA 717.86 USD
53498-104 53498-103
Pipet Aid® Portable Pipetting Device, Drummond Scientific
Pipets Pipet Controllers
This lightweight, battery-powered pipet controller accepts plastic or glass pipets from 1 to 100 ml.
  • Pipet device works 2000 times without recharging
  • Flow rate better than 25 ml at 5 s with 100 ml pipet
  • Cordless and portable pipet controller
  • Filtered, failsafe pipet nosepiece
  • Holster wall bracket provided
With a rechargeable NiMH battery built into its handle, the portable pipet aid can be used continuously for eight hours, pipetting over 2000 times without recharging.
The media dispenser features a locking tissue culture filter that helps eliminate cross contamination due to over pipetting. The nosepiece incorporates a dual hydrophilic/hydrophobic 0.8 µm filter, which provides a barrier to 0.1 µm particles and prevents further use once contaminated by fluid. Work can be resumed soon after the simple filter replacement and nosepiece decontamination.
The battery charges overnight.
Información accesorios: The tissue culture nosepiece kit is also available separately. Additional filters and an outer shell can be ordered separately.
Información para pedidos: This device includes the tissue culture nosepiece kit. The nosepiece kit is complete with housing, gaskets, rubber insert, and four filters. Additional filters may be ordered separately. A holster is included.