VWR® Disposable Micropipets
Supplier: VWR, part of Avantor
VWR®
2-033-020 2-033-025 2-033-044-H 2-033-005 2-033-075 2-033-050 2-033-100 2-033-044 2-033-010
53432-808CS 602.82 USD
53432-808 15401-413 15401-402 53432-706 53432-728 53432-921 53432-740 53432-761 53432-783
VWR® Disposable Micropipets
Pipets Microcapillaries
Borosilicate glass micropipets are made to exacting specifications and are calibrated to contain with permanent fill lines.
  • Color-coded
  • Borosilicate glass fabrication
  • Sizes from 5 to 100 µl
  • Permanent fill lines
The 44.7 µl size is treated with the anticoagulant heparin for clinical applications.
Each canister is marked with bold size designations for quick identification in the laboratory.
Información para pedidos: This item is packed in easy dispensing canisters of 250 micropipets and one 38.1 cm (15") aspirator tube.