Disposable Milk Pipets, Sterile, DWK Life Sciences
Supplier: DWK Life Sciences
56404-11 56400-11
53384-240CS 1288.38 USD
53384-240 53384-192
Disposable Milk Pipets, Sterile, DWK Life Sciences
Pipets Milk Testing Pipets
Disposable, plugged, polystyrene pipet with uniform pulled tip.
Calibrated for the measurement of undiluted and diluted milk, as prescribed in 'Standard Methods for Analysis of Dairy Products,' American Public Health Association.
Envase: Supplied sterile in bags of 25, individually wrapped.