SP Bel-Art Pipette Pump Pipettors, Bel-Art Products, a part of SP
Supplier: Bel-Art
F37896-0000 F37899-0000 F37897-0000 F37898-0000
490005-654EA 43.81 USD
490005-654 490012-358 490012-362 490012-360
SP Bel-Art Pipette Pump Pipettors, Bel-Art Products, a part of SP
Pipets Pipet Controllers
Provides safe, accurate and trouble-free pipetting with simple, one-handed operation.
  • Pipette fits smoothly into its flexible tapered chuck
  • Thumbwheel on side is rotated for precision aspirating or dispensing
  • Plunger may be pressed for quick emptying
  • Resistant to acids and alkalis
Disassemble all parts for cleaning and before autoclaving at 121 °C (250 °F) for 20 minutes; chuck should not be autoclaved.
Información para pedidos: Replacement silicone rubber chuck and washers are available must be ordered separately.