Disposable Milk Pipets, Glass, Sterile, Plugged, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences
72106-11 72106-22
48754-110CS 495.18 USD
48754-110 48754-104
Disposable Milk Pipets, Glass, Sterile, Plugged, Kimble Chase, DWK Life Sciences
Pipets Milk Testing Pipets
Pipets have a uniform pulled tip and are clean, plugged, sterile, and ready for use.
The 1.1 ml size is also suitable for use in the milk phosphatase test.
Certificaciones: Calibrated to meet APHA requirements for making milk dilutions.
Información para pedidos: Packaged in paper/plastic individual wrap and overpacked in convenient dispensing shelf-packs for laboratory use.