SP Bel-Art Fast Release Pipet Pump™ III Pipettors, Bel-Art Products, a part of SP
Supplier: Bel-Art
Pipette Pump™
379040010 379040002 379040025
47751-790EA 55.37 USD
47751-790 47751-788 47751-786
SP Bel-Art Fast Release Pipet Pump™ III Pipettors, Bel-Art Products, a part of SP
Pipets Pipet Controllers
The ergonomic design of Pipet Pump™ III Pipettors enhances comfort, utility, and dispensing precision while helping to reduce the risk of injury from repetitive and awkward pipetting motions.
  • Ergonomic design minimizes user strain
  • Lightweight, one-handed operation
  • Thumb wheel is used to 'blow out' contents of the pipet
The angle of the inserted pipet and its placement in front of the hand allows the arm to remain in a more natural, lowered position unlike other pipettors, which require arm and elbow elevation for many pipetting actions.
A fast-release trigger dispenses the entire pipet contents while an optimally located thumb wheel easily rotates for precision aspirating or dispensing. Volumes as low as one drop at a time may be set, all while using a relaxed grip. The thumb wheel also can be used to 'blow out' the contents of the pipet.
Available in three different pipet volumes (2, 10 and 25 ml), each color-coded for easy identification. A self-stabilizing collar inside the silicone chuck holds either glass or plastic pipets. Resistant to acids, alkalis, and most solvents, the device can be disassembled for cleaning.
Compact design adds only 15.2 cm (6") to the pipet length, allowing use in fume hoods and controlled environment glove boxes.