SP Bel-Art Heavy Duty Rectangular Tanks with Top Flanges, Bel-Art Products, a part of SP
Supplier: Bel-Art
H34096-0000 H34090-0000 H34094-0000 H34085-0000 H34080-0000 H34092-0000 H34082-0000 H34093-0000 H34095-0000
47750-070EA 337.53 USD
47750-070 70680-130 47750-060 47750-072 47750-074 47750-064 13197-372 47750-066 47750-068
SP Bel-Art Heavy Duty Rectangular Tanks with Top Flanges, Bel-Art Products, a part of SP
Tanks Rectangular Tanks
Seamless, rotational-molded polyethylene tanks are lightweight and easy to handle.
  • Polyethylene construction
  • Seamless rotational mold
  • Lightweight
  • Easy to handle
Available with or without a faucet, these tanks are resistant to most acids and all alkalis. All sizes are made with wide top flanges, include covers.
Información para pedidos: Can be supplied with spigots, fittings, dams, partitions etc. Contact your VWR representative for quotations on special sizes and fabrication in polypropylene and PVC.