SP Bel-Art Precisionware® Bottles, Narrow Mouth, Amber, HDPE, Bel-Art Products, a part of SP
Supplier: Bel-Art
SCIENCEWARE®
10624-9015 10624-9007 10624-9016 10624-9008
47748-104CS 716.4 USD
47748-104 47748-100 47748-102 47748-098
SP Bel-Art Precisionware® Bottles, Narrow Mouth, Amber, HDPE, Bel-Art Products, a part of SP
Bottles Plastic Bottles
FDA compliant laboratory/pharmaceutical grade, amber colored bottles are perfect for the storage and sampling of light sensitive materials.
  • Rigid HDPE construction
  • Polypropylene closures
  • Leakproof design
  • Amber coloring provides protection for light-sensitive materials
Made of rigid high-density polyethylene, they have matched polypropylene closures with a leakproof design. They are ideal for packaging and storage of liquids and powders. Bottles 250 ml (8.4 oz.) and larger feature a recycling code molded into the base.
Certificaciones: Bottles comply with U.S. Pharma 23 requirements for light transmission.