Bochem Stainless Steel Lab Support Jacks, BrandTech
Supplier: Bochem
B11120 B11190 B11130 B11122 B11115 B11140 B11180 B11132
47742-856EA 528.99 USD
47742-856 12000-782 47742-854 12000-784 47742-852 47742-862 47742-860 47742-858
Bochem Stainless Steel Lab Support Jacks, BrandTech
Laboratory Jacks
Improved–Jacks now feature thicker scissor mechanism supports for enhanced stability.
  • 304 stainless deck and lift mechanisms for maximum service life
  • Extra-thick scissor mechanism for stability
  • 30x30 cm and 40x40 cm sizes include wrench for easy adjustment
Support jacks are constructed entirely of 304 stainless steel with POM rollers and phenolic knob for long service life in corrosive environments, such as fume hoods. All-roller design provides smooth stable height adjustment. Maxi-lift models provide 30% more height than standard models. Large (30x30cm and 40x40cm) sizes are supplied with a ratcheting wrench for easy height adjustment. Jacks are manufactured according to DIN 12897.
Información para pedidos: Models B11180 and B11190 include ratchet to assist in adjustment.