SP Bel-Art Utility Trays, Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art
F16300-1520 F16300-1015 F16300-2226 80016-0237 F16300-1216 F16300-1818 F16300-2020
47750-864EA 427.34 USD
47750-864 47750-854 47750-856 47750-858 47750-860 47728-018 47750-862
SP Bel-Art Utility Trays, Polypropylene, Bel-Art Products, a part of SP
Trays/Dishes/Pots Laboratory Trays/Dishes
Made from polypropylene, these straight-sided trays offer resistance to temperatures up to 135 °C (274 °F).
  • Polypropylene construction
  • Good temperature resistance
  • Straight-sided design
  • Autoclavable