ProPette™ Electronic Pipette Controller
Supplier: MTC BIO, INC.
P6080-FI P6080-NA P6080-I P6080 P6080-BA P6080-RC
470228-484EA 48.5 USD
470228-480 470228-484 470228-482 470228-478 470228-488 470228-486
ProPette™ Electronic Pipette Controller
Pipets Pipet Controllers
ProPette™ features an ergonomic body design, with computer-balanced weight distribution and low pressure fingertip control buttons that combine to provide a more comfortable pipetting experience.
  • QuickStand™ prevents pipette contamination
  • Variable speed control
  • Cordless and rechargable
  • Provides 8 hours of continuous operation
  • Fits serological pipettes from 1 to 100 ml
  • Ambidexterous
Retractable Quickstand™ allows the instrument to be set down on the bench without the risk of contaminating pipettes.